Assay Details
Gene Symbol: lmnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203867.1 | NP_989198.1 | 1 - 2 | 586 | 64 | |
GenBank mRNA | BC063368.1 | - | 1 - 2 | 586 | 64 | |
CR942583.1 | - | 1 - 2 | 425 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
