Assay Details
Gene Symbol: wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203868.1 | NP_989199.1 | 5 - 6 | 532 | 64 | |
XM_012962846.2 | XP_012818300.1 | 5 - 6 | 582 | 64 | ||
XM_012962847.2 | XP_012818301.1 | 5 - 6 | 486 | 64 | ||
GenBank mRNA | BC063362.1 | - | 5 - 6 | 532 | 64 | |
CR760675.2 | - | 5 - 6 | 336 | 64 | ||
CR926261.1 | - | 5 - 6 | 445 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
