Assay Details
Gene Symbol: nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203874.1 | NP_989205.1 | 9 - 10 | 1019 | 62 | |
XM_012956657.1 | XP_012812111.1 | 9 - 10 | 1189 | 62 | ||
XM_012956658.2 | XP_012812112.1 | 10 - 11 | 1171 | 62 | ||
XM_018091146.1 | XP_017946635.1 | 9 - 10 | 980 | 62 | ||
XM_018091147.1 | XP_017946636.1 | 8 - 9 | 1007 | 62 | ||
GenBank mRNA | BC063353.1 | - | 9 - 10 | 1019 | 62 | |
CR848293.2 | - | 9 - 10 | 950 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
