Assay Details
Gene Symbol: kctd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203908.1 | NP_989239.1 | 4 - 5 | 856 | 74 | |
XM_012960765.2 | XP_012816219.1 | 4 - 5 | 4182 | 74 | ||
XM_018092973.1 | XP_017948462.1 | 4 - 5 | 1126 | 74 | ||
GenBank mRNA | BC063913.1 | - | 4 - 5 | 856 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
