Assay Details
| Gene Symbol: stk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203918.1 | NP_989249.1 | 7 - 8 | 1005 | 55 | |
| GenBank mRNA | AB744231.1 | - | 7 - 8 | 833 | 55 | |
| BC063903.1 | - | 7 - 8 | 1005 | 55 | ||
| CR848251.2 | - | 4 - 5 | 536 | 55 | ||
More Information
Set Membership: |
|

stk4
Genomic Map