Assay Details
Gene Symbol: cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203926.1 | NP_989257.1 | 2 - 3 | 237 | 75 | |
XM_012960766.2 | XP_012816220.1 | 2 - 3 | 188 | 75 | ||
GenBank mRNA | BC063914.1 | - | 2 - 3 | 237 | 75 | |
BC075515.1 | - | 2 - 3 | 276 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
