Assay Details
| Gene Symbol: snai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203936.1 | NP_989267.1 | 1 - 2 | 163 | 91 | |
| GenBank mRNA | BC064223.1 | - | 1 - 2 | 163 | 91 | |
| CR759990.2 | - | 1 - 2 | 154 | 91 | ||
More Information
Set Membership: |
|

snai1
Genomic Map