Assay Details
| Gene Symbol: tubb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203944.1 | NP_989275.1 | 1 - 2 | 170 | 68 | |
| GenBank mRNA | BC064166.1 | - | 1 - 2 | 170 | 68 | |
More Information
Set Membership: |
|

tubb2b
Genomic Map