Assay Details
| Gene Symbol: alg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203952.1 | NP_989283.1 | 3 - 4 | 1198 | 57 | |
| GenBank mRNA | BC064222.1 | - | 3 - 4 | 1198 | 57 | |
More Information
Set Membership: |
|

alg11
Genomic Map