Assay Details
| Gene Symbol: rbbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203954.1 | NP_989285.1 | 5 - 6 | 696 | 67 | |
| GenBank mRNA | BC064219.1 | - | 5 - 6 | 696 | 67 | |
| BC080146.1 | - | 5 - 6 | 735 | 67 | ||
More Information
Set Membership: |
|

rbbp7
Genomic Map