Assay Details
Gene Symbol: adap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203955.1 | NP_989286.1 | 1 - 2 | 215 | 75 | |
XM_018092976.1 | XP_017948465.1 | 1 - 2 | 344 | 75 | ||
XM_018092977.1 | XP_017948466.1 | 1 - 2 | 346 | 75 | ||
XM_018092978.1 | XP_017948467.1 | 1 - 2 | 338 | 75 | ||
GenBank mRNA | BC064211.1 | - | 1 - 2 | 215 | 75 | |
BC075518.1 | - | 1 - 2 | 258 | 75 | ||
CR760308.2 | - | 1 - 2 | 136 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
