Assay Details
Gene Symbol: adap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203955.1 | NP_989286.1 | 3 - 4 | 438 | 88 | |
XM_012960771.2 | XP_012816225.1 | 3 - 4 | 685 | 88 | ||
XM_012960772.2 | XP_012816226.1 | 2 - 3 | 436 | 88 | ||
XM_018092976.1 | XP_017948465.1 | 3 - 4 | 567 | 88 | ||
XM_018092977.1 | XP_017948466.1 | 3 - 4 | 569 | 88 | ||
XM_018092978.1 | XP_017948467.1 | 3 - 4 | 561 | 88 | ||
XM_018092979.1 | XP_017948468.1 | 2 - 3 | 1559 | 88 | ||
GenBank mRNA | BC064211.1 | - | 3 - 4 | 438 | 88 | |
BC075518.1 | - | 3 - 4 | 481 | 88 | ||
CR760308.2 | - | 3 - 4 | 359 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
