Assay Details
Gene Symbol: adap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203955.1 | NP_989286.1 | 7 - 8 | 855 | 100 | |
XM_012960771.2 | XP_012816225.1 | 7 - 8 | 1102 | 100 | ||
XM_012960772.2 | XP_012816226.1 | 6 - 7 | 853 | 100 | ||
XM_018092977.1 | XP_017948466.1 | 6 - 7 | 839 | 100 | ||
XM_018092978.1 | XP_017948467.1 | 7 - 8 | 978 | 100 | ||
XM_018092979.1 | XP_017948468.1 | 6 - 7 | 1976 | 100 | ||
GenBank mRNA | BC064211.1 | - | 7 - 8 | 855 | 100 | |
BC075518.1 | - | 7 - 8 | 898 | 100 | ||
CR760308.2 | - | 7 - 8 | 776 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
