Assay Details
Gene Symbol: adap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203955.1 | NP_989286.1 | 9 - 10 | 996 | 95 | |
XM_012960771.2 | XP_012816225.1 | 9 - 10 | 1243 | 95 | ||
XM_012960772.2 | XP_012816226.1 | 8 - 9 | 994 | 95 | ||
XM_018092976.1 | XP_017948465.1 | 8 - 9 | 1041 | 95 | ||
XM_018092977.1 | XP_017948466.1 | 8 - 9 | 980 | 95 | ||
XM_018092979.1 | XP_017948468.1 | 8 - 9 | 2117 | 95 | ||
GenBank mRNA | BC064211.1 | - | 9 - 10 | 996 | 95 | |
CR760308.2 | - | 9 - 10 | 917 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
