Assay Details
Gene Symbol: mrpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203957.1 | NP_989288.1 | 2 - 3 | 338 | 60 | |
GenBank mRNA | BC064179.1 | - | 2 - 3 | 338 | 60 | |
CR761345.2 | - | 2 - 3 | 230 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
