Assay Details
| Gene Symbol: ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203974.1 | NP_989305.1 | 2 - 3 | 337 | 73 | |
| XM_012970266.2 | XP_012825720.1 | 2 - 3 | 484 | 73 | ||
| GenBank mRNA | BC064216.1 | - | 2 - 3 | 337 | 73 | |
| BC082739.1 | - | 2 - 3 | 339 | 73 | ||
| CR762319.2 | - | 1 - 2 | 216 | 73 | ||
More Information
Set Membership: |
|

ube2e3
Genomic Map