Assay Details
Gene Symbol: ube2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203974.1 | NP_989305.1 | 5 - 6 | 667 | 76 | |
XM_012970266.2 | XP_012825720.1 | 5 - 6 | 814 | 76 | ||
GenBank mRNA | BC064216.1 | - | 5 - 6 | 667 | 76 | |
BC082739.1 | - | 5 - 6 | 669 | 76 | ||
CR762319.2 | - | 4 - 5 | 546 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
