Assay Details
| Gene Symbol: uqcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203977.1 | NP_989308.1 | 3 - 4 | 247 | 60 | |
| GenBank mRNA | BC064265.1 | - | 3 - 4 | 247 | 60 | |
| BC086302.1 | - | 3 - 4 | 284 | 60 | ||
| CR761317.2 | - | 3 - 4 | 274 | 60 | ||
More Information
Set Membership: |
|

uqcc1
Genomic Map