Assay Details
| Gene Symbol: pdia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203998.1 | NP_989329.1 | 12 - 13 | 1437 | 85 | |
| GenBank mRNA | BC064163.1 | - | 12 - 13 | 1437 | 85 | |
| CR760291.2 | - | 12 - 13 | 1438 | 85 | ||
More Information
Set Membership: |
|

pdia3
Genomic Map