Assay Details
| Gene Symbol: atp6v0d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204031.1 | NP_989362.1 | 6 - 7 | 847 | 77 | |
| GenBank mRNA | BC064198.1 | - | 6 - 7 | 847 | 77 | |
| CR760271.2 | - | 6 - 7 | 847 | 77 | ||
More Information
Set Membership: |
|

atp6v0d2
Genomic Map