Assay Details
| Gene Symbol: ckmt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204038.1 | NP_989369.1 | 2 - 3 | 191 | 74 | |
| XM_018092012.1 | XP_017947501.1 | 2 - 3 | 190 | 74 | ||
| GenBank mRNA | BC064224.1 | - | 2 - 3 | 191 | 74 | |
| BC076879.1 | - | 2 - 3 | 205 | 74 | ||
| CR760861.2 | - | 2 - 3 | 206 | 74 | ||
More Information
Set Membership: |
|

ckmt1b
Genomic Map