Assay Details
| Gene Symbol: ckmt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204038.1 | NP_989369.1 | 5 - 6 | 705 | 101 | |
| XM_018092012.1 | XP_017947501.1 | 5 - 6 | 704 | 101 | ||
| GenBank mRNA | BC064224.1 | - | 5 - 6 | 705 | 101 | |
| BC076879.1 | - | 5 - 6 | 719 | 101 | ||
| CR760861.2 | - | 5 - 6 | 720 | 101 | ||
More Information
Set Membership: |
|

ckmt1b
Genomic Map