Assay Details
Gene Symbol: ckmt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204038.1 | NP_989369.1 | 6 - 7 | 796 | 84 | |
XM_018092012.1 | XP_017947501.1 | 6 - 7 | 795 | 84 | ||
GenBank mRNA | BC064224.1 | - | 6 - 7 | 796 | 84 | |
BC076879.1 | - | 6 - 7 | 810 | 84 | ||
CR760861.2 | - | 6 - 7 | 811 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
