Assay Details
Gene Symbol: glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204039.2 | NP_989370.2 | 3 - 4 | 276 | 59 | |
XM_018091151.1 | XP_017946640.1 | 3 - 4 | 353 | 59 | ||
GenBank mRNA | BC064201.1 | - | 3 - 4 | 276 | 59 | |
CR761634.2 | - | 3 - 4 | 498 | 59 | ||
CT485719.1 | - | 3 - 4 | 526 | 59 | ||
CX931067.1 | - | 3 - 4 | 255 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
