Assay Details
| Gene Symbol: kdelr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204050.1 | NP_989381.1 | 3 - 4 | 432 | 86 | |
| GenBank mRNA | BC064720.1 | - | 3 - 4 | 432 | 86 | |
| CT030103.2 | - | 3 - 4 | 417 | 86 | ||
More Information
Set Membership: |
|

kdelr1
Genomic Map