Assay Details
Gene Symbol: anp32c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204073.1 | NP_989404.1 | 2 - 3 | 232 | 68 | |
XM_018091892.1 | XP_017947381.1 | 2 - 3 | 292 | 68 | ||
GenBank mRNA | BC064864.1 | - | 2 - 3 | 232 | 68 | |
CR855787.2 | - | 2 - 3 | 227 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
