Assay Details
Gene Symbol: cdx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204085.1 | NP_989416.1 | 1 - 2 | 547 | 96 | |
GenBank mRNA | AF417198.1 | - | 1 - 2 | 547 | 96 | |
BC171326.1 | - | 1 - 2 | 544 | 96 | ||
BC171327.1 | - | 1 - 2 | 544 | 96 | ||
CR760338.2 | - | 1 - 2 | 495 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
