Assay Details
Gene Symbol: foxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204092.1 | NP_989423.1 | 1 - 2 | 100 | 68 | |
GenBank mRNA | AY260736.1 | - | 1 - 2 | 100 | 68 | |
BC088590.1 | - | 1 - 2 | 56 | 68 | ||
BC170757.1 | - | 1 - 2 | 94 | 68 | ||
BC171324.1 | - | 1 - 2 | 94 | 68 | ||
CR848398.2 | - | 1 - 2 | 100 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
