Assay Details
| Gene Symbol: ensa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213680.1 | NP_998845.1 | 2 - 3 | 475 | 77 | |
| GenBank mRNA | BC067944.1 | - | 2 - 3 | 475 | 77 | |
| CR760899.2 | - | 2 - 3 | 242 | 77 | ||
More Information
Set Membership: |
|

ensa
Genomic Map