Assay Details
| Gene Symbol: ensa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213680.1 | NP_998845.1 | 3 - 4 | 652 | 75 | |
| GenBank mRNA | BC067944.1 | - | 3 - 4 | 652 | 75 | |
| CR760899.2 | - | 3 - 4 | 419 | 75 | ||
More Information
Set Membership: |
|

ensa
Genomic Map