Assay Details
Gene Symbol: herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213681.2 | NP_998846.1 | 6 - 7 | 887 | 67 | |
XM_012960788.2 | XP_012816242.1 | 6 - 7 | 934 | 67 | ||
XM_018092980.1 | XP_017948469.1 | 5 - 6 | 856 | 67 | ||
XM_018092981.1 | XP_017948470.1 | 5 - 6 | 833 | 67 | ||
GenBank mRNA | BC067925.2 | - | 6 - 7 | 887 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
