Assay Details
| Gene Symbol: rpl26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005104.2 | NP_001005104.1 | 2 - 3 | 209 | 67 | |
| XM_012958854.2 | XP_012814308.1 | 2 - 3 | 250 | 67 | ||
| GenBank mRNA | BC077038.1 | - | 2 - 3 | 210 | 67 | |
| BC161328.1 | - | 2 - 3 | 202 | 67 | ||
| CR761946.2 | - | 1 - 2 | 177 | 67 | ||
More Information
Set Membership: |
|

rpl26
Genomic Map