Assay Details
| Gene Symbol: atp5mc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005112.2 | NP_001005112.1 | 389 | 78 | ||
| GenBank mRNA | BC077049.1 | - | 394 | 78 | ||
| CR761581.2 | - | 319 | 78 | |||
More Information
Set Membership: |
|

atp5mc1
Genomic Map