Assay Details
| Gene Symbol: rps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005113.2 | NP_001005113.1 | 375 | 74 | ||
| GenBank mRNA | BC077050.1 | - | 376 | 74 | ||
| CR760469.2 | - | 358 | 74 | |||
More Information
Set Membership: |
|

rps11
Genomic Map