Assay Details
Gene Symbol: eif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005114.1 | NP_001005114.1 | 3 - 4 | 396 | 91 | |
GenBank mRNA | BC077051.1 | - | 3 - 4 | 396 | 91 | |
BC161044.1 | - | 3 - 4 | 369 | 91 | ||
CR761201.2 | - | 3 - 4 | 300 | 91 | ||
CT030083.2 | - | 3 - 4 | 362 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
