Assay Details
Gene Symbol: ndufab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005125.1 | NP_001005125.1 | 1 - 2 | 182 | 62 | |
GenBank mRNA | BC077661.1 | - | 1 - 2 | 182 | 62 | |
CR760725.2 | - | 1 - 2 | 160 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
