Assay Details
| Gene Symbol: apoa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005127.1 | NP_001005127.1 | 3 - 4 | 246 | 73 | |
| XM_012966446.2 | XP_012821900.1 | 3 - 4 | 302 | 73 | ||
| GenBank mRNA | BC077663.1 | - | 3 - 4 | 246 | 73 | |
| CR760756.2 | - | 2 - 3 | 230 | 73 | ||
More Information
Set Membership: |
|

apoa1
Genomic Map