Assay Details
| Gene Symbol: ap3s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005131.1 | NP_001005131.1 | 4 - 5 | 520 | 81 | |
| XM_012960204.2 | XP_012815658.1 | 4 - 5 | 639 | 81 | ||
| GenBank mRNA | BC077669.1 | - | 4 - 5 | 520 | 81 | |
| CR760027.2 | - | 4 - 5 | 383 | 81 | ||
More Information
Set Membership: |
|

ap3s1
Genomic Map