Assay Details
| Gene Symbol: rpl35a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005134.2 | NP_001005134.1 | 3 - 4 | 231 | 69 | |
| XM_012962901.1 | XP_012818355.1 | 3 - 4 | 680 | 69 | ||
| XM_012962902.1 | XP_012818356.1 | 3 - 4 | 695 | 69 | ||
| GenBank mRNA | BC077673.1 | - | 3 - 4 | 233 | 69 | |
| CR761941.2 | - | 2 - 3 | 195 | 69 | ||
More Information
Set Membership: |
|

rpl35a
Genomic Map