Assay Details
Gene Symbol: rpl37a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005137.1 | NP_001005137.1 | 2 - 3 | 150 | 69 | |
GenBank mRNA | BC077677.1 | - | 2 - 3 | 150 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
