Assay Details
Gene Symbol: nme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005140.2 | NP_001005140.1 | 3 - 4 | 268 | 89 | |
XM_012954135.1 | XP_012809589.1 | 2 - 3 | 236 | 89 | ||
GenBank mRNA | BC077684.1 | - | 3 - 4 | 269 | 89 | |
CR760723.2 | - | 2 - 3 | 237 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
