Assay Details
Gene Symbol: snrpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005147.2 | NP_001005147.1 | 4 - 5 | 264 | 108 | |
GenBank mRNA | BC077697.1 | - | 4 - 5 | 266 | 108 | |
BC159375.1 | - | 4 - 5 | 283 | 108 | ||
CR761996.2 | - | 4 - 5 | 220 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
