Assay Details
Gene Symbol: fnbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005148.1 | NP_001005148.1 | 2 - 3 | 233 | 81 | |
XM_012960886.2 | XP_012816340.1 | 2 - 3 | 365 | 81 | ||
XM_018093007.1 | XP_017948496.1 | 2 - 3 | 365 | 81 | ||
XM_018093008.1 | XP_017948497.1 | 2 - 3 | 365 | 81 | ||
GenBank mRNA | AY640054.1 | - | 2 - 3 | 139 | 81 | |
BC080954.1 | - | 2 - 3 | 233 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
