Assay Details
Gene Symbol: hdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005432.1 | NP_001005432.1 | 1 - 2 | 243 | 69 | |
GenBank mRNA | BC074509.1 | - | 1 - 2 | 243 | 69 | |
CR926395.2 | - | 1 - 2 | 199 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
