Assay Details
| Gene Symbol: poldip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005439.1 | NP_001005439.1 | 6 - 7 | 881 | 62 | |
| XM_012960800.2 | XP_012816254.1 | 6 - 7 | 1006 | 62 | ||
| GenBank mRNA | BC074542.1 | - | 6 - 7 | 881 | 62 | |
| CR855797.2 | - | 6 - 7 | 918 | 62 | ||
More Information
Set Membership: |
|

poldip3
Genomic Map