Assay Details
Gene Symbol: ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005452.1 | NP_001005452.1 | 4 - 5 | 832 | 75 | |
GenBank mRNA | BC074566.1 | - | 4 - 5 | 832 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
