Assay Details
| Gene Symbol: adsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005457.1 | NP_001005457.1 | 10 - 11 | 1211 | 62 | |
| GenBank mRNA | BC074577.1 | - | 10 - 11 | 1211 | 62 | |
| CR762315.2 | - | 10 - 11 | 1199 | 62 | ||
More Information
Set Membership: |
|

adsl
Genomic Map