Assay Details
| Gene Symbol: pisd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005623.1 | NP_001005623.1 | 4 - 5 | 474 | 68 | |
| XM_012970288.2 | XP_012825742.1 | 4 - 5 | 573 | 68 | ||
| GenBank mRNA | BC074595.1 | - | 4 - 5 | 474 | 68 | |
More Information
Set Membership: |
|

pisd
Genomic Map