Assay Details
Gene Symbol: eif2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005630.1 | NP_001005630.1 | 6 - 7 | 757 | 63 | |
GenBank mRNA | BC074615.1 | - | 6 - 7 | 757 | 63 | |
CR761726.2 | - | 6 - 7 | 748 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
