Assay Details
Gene Symbol: grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005632.1 | NP_001005632.1 | 8 - 9 | 1050 | 71 | |
XM_012956715.2 | XP_012812169.1 | 7 - 8 | 966 | 71 | ||
GenBank mRNA | BC074617.1 | - | 8 - 9 | 1050 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
